missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 0.5M buffer soln., pH 7.5, Thermo Scientific™

Catalog No. AAJ62672AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62672AE 100 mL
AAJ62672AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62672AE Supplier Thermo Scientific Chemicals Supplier No. J62672AE
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Color Colorless
Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Tricine buffer soln.
Hazard Category H335
TSCA Yes
Recommended Storage Keep cold