missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 0.5M buffer soln., pH 6.5, Thermo Scientific™

$100.62 - $165.22

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60107AE
View Documents
Thermo Scientific Chemicals
J60107AE
100 mL
Each for $100.62
Only null left
 
AAJ60107AK
View Documents
Thermo Scientific Chemicals
J60107AK
250 mL
Each for $165.22
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4-1-5704
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO
C6H13NO5
MFCD00004277
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid

Specifications

4-1-5704
0.5 M
Liquid
C6H13NO5
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
Tricine buffer soln.
Colorless
6.5
100 mL
MFCD00004277
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
Videos
Safety and Handling

Safety and Handling

H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'