missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine, 0.5M buffer soln., pH 6.5, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Colorless |
| Concentration | 0.5 M |
| pH | 6.5 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Chemical Name or Material | Tricine buffer soln. |
Safety and Handling
| Hazard Category | H335 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |