Learn More
alpha,alpha,alpha-Trichlorotoluene, 98%
CAS: 98-07-7 | C7H5Cl3 | 195.47 g/mol
Supplier: Thermo Scientific Chemicals 164530010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| α, α, α-Trichlorotoluene | |
| 98-07-7 | |
| 100.0 | |
| Yellow | |
| 219°C to 223°C | |
| Authentic | |
| Glass bottle | |
| 1.5570 to 1.559 | |
| MFCD00000786 | |
| 05, 300 | |
| 15, 1112 | |
| Solubility in water: hydrolysis. Other solubilities: soluble in alcohol,benzene,ether and,most organic solvents | |
| ClC(Cl)(Cl)C1=CC=CC=C1 | |
| 195.47 | |
| CHEBI:82274 | |
| 98% |
| 98% | |
| 97.5 | |
| -7.5°C | |
| 1.3800g/mL | |
| 97°C | |
| 98% | |
| C7H5Cl3 | |
| C6H5CCl3 | |
| 1 L | |
| 1.38 | |
| benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane | |
| XEMRAKSQROQPBR-UHFFFAOYSA-N | |
| trichloromethylbenzene | |
| 7367 | |
| 195.48 | |
| Liquid |
Chemical Identifiers
| 98-07-7 | |
| 195.47 | |
| XEMRAKSQROQPBR-UHFFFAOYSA-N | |
| 7367 | |
| trichloromethylbenzene |
| C7H5Cl3 | |
| MFCD00000786 | |
| benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane | |
| CHEBI:82274 | |
| ClC(Cl)(Cl)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Toxic if inhaled.
May cause cancer.
May cause respiratory irritation.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-634-5
RTECSNumber : XT9275000
TSCA : TSCA
RUO – Research Use Only