missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trichlorotoluene, 98%

Catalog No. p-202903
Change view
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164530050 5 mL Glass bottle
AC164530010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164530050 Supplier Thermo Scientific Chemicals Supplier No. 164530050
Only null left

CAS: 98-07-7 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-07-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000786
InChI Key XEMRAKSQROQPBR-UHFFFAOYSA-N
Synonym benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
PubChem CID 7367
ChEBI CHEBI:82274
IUPAC Name trichloromethylbenzene
SMILES ClC(Cl)(Cl)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -7.5°C
Color Yellow
Density 1.3800g/mL
Boiling Point 219°C to 223°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5570 to 1.559
Linear Formula C6H5CCl3
Quantity 5 mL
Beilstein 05, 300
Specific Gravity 1.38
Merck Index 15, 1112
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in alcohol,benzene,ether and,most organic solvents
Formula Weight 195.48
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α, α, α-Trichlorotoluene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Toxic if inhaled.
May cause cancer.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-634-5
RTECSNumber XT9275000
TSCA TSCA

RUO – Research Use Only