missing translation for 'onlineSavingsMsg'
Learn More

Trichloro(methyl)silane 98.0+%, TCI America™

Supplier:  TCI America M0450100G

 View more versions of this product

Catalog No. M0450100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trichloro(methyl)silane
-90°C
66°C
MFCD00000481
1250
JLUFWMXJHAVVNN-UHFFFAOYSA-N
trichloro(methyl)silane
6399
≥98.0% (GC,T)
75-79-6
Colorless
CH3Cl3Si
100 g
methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
C[Si](Cl)(Cl)Cl
149.47
149.47
Liquid

Chemical Identifiers

75-79-6
149.47
JLUFWMXJHAVVNN-UHFFFAOYSA-N
6399
C[Si](Cl)(Cl)Cl
CH3Cl3Si
MFCD00000481
methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
trichloro(methyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2041

RTECSNumber : VV4550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions