accessibility menu, dialog, popup

UserName

CH3SiClClCl

CAS RN 75-79-6

IUPAC Name: trichloro(methyl)silane
Synonyms: methyltrichlorosilane
Molecular Weight (g/mol): 149.47
Molecular Formula: CH3Cl3Si
InChi Key: JLUFWMXJHAVVNN-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (5)