missing translation for 'onlineSavingsMsg'
Learn More

Trichloro(methyl)silane 98.0+%, TCI America™

Catalog No. p-7132178
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045025G 25 g
M0450100G 100 g
M0450500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M045025G Supplier TCI America Supplier No. M045025G

Chemical Identifiers

CAS 75-79-6
Molecular Formula CH3Cl3Si
Molecular Weight (g/mol) 149.47
MDL Number MFCD00000481
InChI Key JLUFWMXJHAVVNN-UHFFFAOYSA-N
Synonym methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
PubChem CID 6399
IUPAC Name trichloro(methyl)silane
SMILES C[Si](Cl)(Cl)Cl

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 66°C
Quantity 25 g
UN Number 1250
Formula Weight 149.47
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Trichloro(methyl)silane
EINECSNumber (2)-2041
RTECSNumber VV4550000
TSCA Yes