missing translation for 'onlineSavingsMsg'
Learn More

Trichloro[3-(pentafluorophenyl)propyl]silane 98.0+%, TCI America™

Catalog No. T30301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30301G 1 g
T30305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30301G Supplier TCI America Supplier No. T30301G

Chemical Identifiers

CAS 78900-02-4
Molecular Formula C9H6Cl3F5Si
Molecular Weight (g/mol) 343.574
MDL Number MFCD01074474
InChI Key PGOAAUBOHVGLCX-UHFFFAOYSA-N
PubChem CID 2760298
IUPAC Name trichloro-[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
SMILES C(CC1=C(C(=C(C(=C1F)F)F)F)F)C[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Quantity 1 g
UN Number 2987
Formula Weight 343.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro[3-(pentafluorophenyl)propyl]silane