accessibility menu, dialog, popup

UserName

FFFFFSiClClCl

CAS RN 78900-02-4

IUPAC Name: trichloro[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
Synonyms: trichloro[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
Molecular Weight (g/mol): 343.57
Molecular Formula: C9H6Cl3F5Si
InChi Key: PGOAAUBOHVGLCX-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(CCC[Si](Cl)(Cl)Cl)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results