missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl(ethyl)phosphonium Diethyl Phosphate 96.0+%, TCI America™
Supplier: TCI America T364125G
Specifications
| Tributyl(ethyl)phosphonium Diethyl Phosphate | |
| Yellow | |
| MFCD17018975 | |
| RDCTYOLAEUHRGG-UHFFFAOYSA-M | |
| diethyl phosphate;tributyl(ethyl)phosphanium | |
| 60196373 | |
| ≥96.0% (T) |
| 20445-94-7 | |
| C18H42O4P2 | |
| 25 g | |
| CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC | |
| 384.478 | |
| 384.48 | |
| Liquid |
Chemical Identifiers
| 20445-94-7 | |
| 384.478 | |
| RDCTYOLAEUHRGG-UHFFFAOYSA-M | |
| diethyl phosphate;tributyl(ethyl)phosphanium |
| C18H42O4P2 | |
| MFCD17018975 | |
| 60196373 | |
| CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC |
Safety and Handling
TSCA : No