accessibility menu, dialog, popup

UserName

CAS RN 20445-94-7

H3CPH3CH3CH3CCH3OPOOOH3C

CAS RN 20445-94-7

IUPAC Name: tributyl(ethyl)phosphanium diethyl phosphate
Synonyms: tributyl(ethyl)phosphanium diethyl phosphate
Molecular Weight (g/mol): 384.48
Molecular Formula: C18H42O4P2
InChi Key: RDCTYOLAEUHRGG-UHFFFAOYSA-M
SMILES: CCOP([O-])(=O)OCC.CCCC[P+](CC)(CCCC)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results