missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl(ethyl)phosphonium Diethyl Phosphate 96.0+%, TCI America™
$149.13 - $507.02
Chemical Identifiers
| CAS | 20445-94-7 |
|---|---|
| Molecular Formula | C18H42O4P2 |
| Molecular Weight (g/mol) | 384.478 |
| MDL Number | MFCD17018975 |
| InChI Key | RDCTYOLAEUHRGG-UHFFFAOYSA-M |
| PubChem CID | 60196373 |
| IUPAC Name | diethyl phosphate;tributyl(ethyl)phosphanium |
| SMILES | CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC |
Chemical Identifiers
| 20445-94-7 | |
| 384.478 | |
| RDCTYOLAEUHRGG-UHFFFAOYSA-M | |
| diethyl phosphate;tributyl(ethyl)phosphanium |
| C18H42O4P2 | |
| MFCD17018975 | |
| 60196373 | |
| CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC |
Specifications
| 20445-94-7 | |
| C18H42O4P2 | |
| 5 g | |
| CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC | |
| 384.478 | |
| 384.48 | |
| Liquid |
| Yellow | |
| MFCD17018975 | |
| RDCTYOLAEUHRGG-UHFFFAOYSA-M | |
| diethyl phosphate;tributyl(ethyl)phosphanium | |
| 60196373 | |
| ≥96.0% (T) | |
| Tributyl(ethyl)phosphonium Diethyl Phosphate |
Safety and Handling
TSCA : No