missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-o-tolyl Borate 98.0+%, TCI America™
$231.14 - $674.79
Chemical Identifiers
| CAS | 2665-12-5 |
|---|---|
| Molecular Formula | C21H21BO3 |
| Molecular Weight (g/mol) | 332.21 |
| MDL Number | MFCD00059311 |
| InChI Key | RTMBXAOPKJNOGZ-UHFFFAOYSA-N |
| Synonym | Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester |
| PubChem CID | 75875 |
| IUPAC Name | tris(2-methylphenyl) borate |
| SMILES | B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C |
Chemical Identifiers
| 2665-12-5 | |
| 332.21 | |
| RTMBXAOPKJNOGZ-UHFFFAOYSA-N | |
| 75875 | |
| B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C |
| C21H21BO3 | |
| MFCD00059311 | |
| Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester | |
| tris(2-methylphenyl) borate |
Specifications
| 2665-12-5 | |
| 386°C | |
| MFCD00059311 | |
| Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester | |
| B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C | |
| 332.21 | |
| 332.21 | |
| Liquid |
| Yellow | |
| C21H21BO3 | |
| 5 mL | |
| RTMBXAOPKJNOGZ-UHFFFAOYSA-N | |
| tris(2-methylphenyl) borate | |
| 75875 | |
| ≥98.0% (T) | |
| Tri-o-tolyl Borate |
Safety and Handling
RTECSNumber : ED4960000
TSCA : No