missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-o-tolyl Borate 98.0+%, TCI America™
Supplier: TCI America B051925ML
Specifications
| Tri-o-tolyl Borate | |
| Yellow | |
| C21H21BO3 | |
| 25 mL | |
| RTMBXAOPKJNOGZ-UHFFFAOYSA-N | |
| tris(2-methylphenyl) borate | |
| 75875 | |
| ≥98.0% (T) |
| 2665-12-5 | |
| 386°C | |
| MFCD00059311 | |
| Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester | |
| B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C | |
| 332.21 | |
| 332.21 | |
| Liquid |
Chemical Identifiers
| 2665-12-5 | |
| 332.21 | |
| RTMBXAOPKJNOGZ-UHFFFAOYSA-N | |
| 75875 | |
| B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C |
| C21H21BO3 | |
| MFCD00059311 | |
| Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester | |
| tris(2-methylphenyl) borate |
Safety and Handling
RTECSNumber : ED4960000
TSCA : No