Learn More
Tri(m-tolyl)phosphine, 98+%
CAS: 6224-63-1 | C21H21P | 304.373 g/mol
Supplier: Thermo Scientific Chemicals A1537206
Description
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
Specifications
| Tri(m-tolyl)phosphine | |
| 99°C to 101°C | |
| MFCD00008532 | |
| 658863 | |
| tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane | |
| LFNXCUNDYSYVJY-UHFFFAOYSA-N | |
| tris(3-methylphenyl)phosphane | |
| 80362 | |
| ≥98% |
| 6224-63-1 | |
| C21H21P | |
| 5 g | |
| Air Sensitive | |
| Insoluble in water. | |
| CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C | |
| 304.373 | |
| 304.37 |
Chemical Identifiers
| 6224-63-1 | |
| 304.373 | |
| LFNXCUNDYSYVJY-UHFFFAOYSA-N | |
| 80362 | |
| CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C |
| C21H21P | |
| MFCD00008532 | |
| tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane | |
| tris(3-methylphenyl)phosphane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-312-4
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only