missing translation for 'onlineSavingsMsg'
Learn More

Tri(m-tolyl)phosphine, 98+%

Catalog No. p-7022544
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1537203 1 g
AAA1537206 5 g
AAA1537214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1537203 Supplier Thermo Scientific Chemicals Supplier No. A1537203

CAS: 6224-63-1 | C21H21P | 304.373 g/mol

Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.

Chemical Identifiers

CAS 6224-63-1
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008532
InChI Key LFNXCUNDYSYVJY-UHFFFAOYSA-N
Synonym tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane
PubChem CID 80362
IUPAC Name tris(3-methylphenyl)phosphane
SMILES CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C

Specifications

Melting Point 99°C to 101°C
Quantity 1 g
Beilstein 658863
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 304.37
Percent Purity ≥98%
Chemical Name or Material Tri(m-tolyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-312-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only