missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Trehalose, Dihydrate
Non-reducing disaccharide which yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro.
Supplier: MilliporeSigma™ 6256251KG
| Quantity | 1 kg |
|---|
Description
- CAS Number: 6138-23-4
- Form: Solid
- Synonyms: 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
- Formula Weight: 378.3g/mol
Chemical Identifiers
| 6138-23-4 | |
| 378.33 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
| C12H26O13 | |
| MFCD00071594 | |
| 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
| 6138-23-4 | |
| MFCD00071594 | |
| 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 378.33 |
| C12H26O13 | |
| 1 kg | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate | |
| 378.3g/mol |