missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transport Medium

Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination

$272.90 - $691.48

Specifications

Certifications/Compliance Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
For Use With (Application) Research, Manufacturing Applications
Quantity 6/Cs.
Storage Requirements 2°C to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
MT25500CI
View Documents Promotion Details
Corning™
25500CI
100 mL
Case of 6 Each for $272.90
Only null left
 
MT25500CV
View Documents Promotion Details
Corning™
25500CV
500 mL
Case of 6 Each for $691.48
Only null left
 
Description

Description

  • This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
  • For research and further manufacturing purposes only
  • Not for direct use in humans or animals.
  • Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
  • Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
Specifications

Specifications

Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
Research, Manufacturing Applications
6/Cs.
2°C to 8°C
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents