missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Transport Medium
Description
- This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
- For research and further manufacturing purposes only
- Not for direct use in humans or animals.
- Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
- Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
Specifications
Specifications
| Certifications/Compliance | Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines |
| For Use With (Application) | Research, Manufacturing Applications |
| Quantity | 6/Cs. |
| Storage Requirements | 2°C to 8°C |
| Volume (Metric) | 100 mL |