missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transport Medium

Catalog No. MT25500CV
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
MT25500CV 500 mL
MT25500CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT25500CV Supplier Corning™ Supplier No. 25500CV

Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination

  • This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
  • For research and further manufacturing purposes only
  • Not for direct use in humans or animals.
  • Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
  • Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B

Specifications

Certifications/Compliance Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
For Use With (Application) Research, Manufacturing Applications
Quantity 6/Cs.
Storage Requirements 2°C to 8°C
Volume (Metric) 500 mL