missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl 98.0+%, TCI America™

$59.65 - $214.37

Chemical Identifiers

CAS 279246-65-0
Molecular Formula C18H32
Molecular Weight (g/mol) 248.45
MDL Number MFCD12761279
InChI Key IASLDLGGHXYCEO-UHFFFAOYSA-N
PubChem CID 22290094
IUPAC Name 4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
SMILES CCCC1CCC(CC1)C1CCC(CC1)C=CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P23711G
View Documents
TCI America
P23711G
1 g N/A
Only null left
 
P23715G
View Documents
TCI America
P23715G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

279246-65-0
248.45
IASLDLGGHXYCEO-UHFFFAOYSA-N
4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
C18H32
MFCD12761279
22290094
CCCC1CCC(CC1)C1CCC(CC1)C=CC

Specifications

279246-65-0
White
MFCD12761279
IASLDLGGHXYCEO-UHFFFAOYSA-N
4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
22290094
≥98.0% (GC)
trans,trans-4-[(E)-1-Propenyl]-4′-propylbicyclohexyl
45°C
C18H32
1 g
CCCC1CCC(CC1)C1CCC(CC1)C=CC
248.45
248.45
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1972

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'