missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl 98.0+%, TCI America™

Supplier:  TCI America P23715G

 View more versions of this product

Catalog No. P23715G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

trans,trans-4-[(E)-1-Propenyl]-4′-propylbicyclohexyl
45°C
C18H32
5 g
CCCC1CCC(CC1)C1CCC(CC1)C=CC
248.45
248.45
Crystalline Lumps
279246-65-0
White
MFCD12761279
IASLDLGGHXYCEO-UHFFFAOYSA-N
4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
22290094
≥98.0% (GC)

Chemical Identifiers

279246-65-0
248.45
IASLDLGGHXYCEO-UHFFFAOYSA-N
4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
C18H32
MFCD12761279
22290094
CCCC1CCC(CC1)C1CCC(CC1)C=CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1972

TSCA : No

SDS
Documents

Documents

Promotions

Promotions