missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl 98.0+%, TCI America™
Supplier: TCI America P23711G
Specifications
| trans,trans-4-[(E)-1-Propenyl]-4′-propylbicyclohexyl | |
| 45°C | |
| C18H32 | |
| 1 g | |
| CCCC1CCC(CC1)C1CCC(CC1)C=CC | |
| 248.45 | |
| 248.45 | |
| Crystalline Lumps |
| 279246-65-0 | |
| White | |
| MFCD12761279 | |
| IASLDLGGHXYCEO-UHFFFAOYSA-N | |
| 4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane) | |
| 22290094 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 279246-65-0 | |
| 248.45 | |
| IASLDLGGHXYCEO-UHFFFAOYSA-N | |
| 4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane) |
| C18H32 | |
| MFCD12761279 | |
| 22290094 | |
| CCCC1CCC(CC1)C1CCC(CC1)C=CC |
Safety and Handling
EINECSNumber : (4)-1972
TSCA : No