missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™

Catalog No. F11021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11021G 1 g
F11025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11021G Supplier TCI America Supplier No. F11021G
Only null left

Chemical Identifiers

CAS 82832-27-7
Molecular Formula C21H31F
Molecular Weight (g/mol) 302.477
MDL Number MFCD11053420
InChI Key IKODGORUTZMKPL-UHFFFAOYSA-N
PubChem CID 578934
IUPAC Name 1-fluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMILES CCCC1CCC(CC1)C2CCC(CC2)C3=CC=C(C=C3)F

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 1 g
Formula Weight 302.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(4-Fluorophenyl)-4′-propylbicyclohexyl