accessibility menu, dialog, popup

UserName

CAS RN 82832-27-7

CH3F

CAS RN 82832-27-7

IUPAC Name: 4-(4-fluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Synonyms: 4-(4-fluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 302.48
Molecular Formula: C21H31F
InChi Key: IKODGORUTZMKPL-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results