missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4'-(3-Butenyl)-4-(3,4-difluorophenyl)bicyclohexyl 98.0+%, TCI America™

Supplier:  TCI America B49165G

 View more versions of this product

Catalog No. B49165G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

trans,trans-4′-(3-Butenyl)-4-(3,4-difluorophenyl)bicyclohexyl
43°C
C22H30F2
5 g
FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(CCC=C)CC1
332.48
332.48
Crystalline Powder
155266-68-5
White
MFCD11053352
UMULWEQZNFZORL-UHFFFAOYSA-N
4-(but-3-en-1-yl)-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
23509154
≥98.0% (GC)

Chemical Identifiers

155266-68-5
332.48
UMULWEQZNFZORL-UHFFFAOYSA-N
4-(but-3-en-1-yl)-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
C22H30F2
MFCD11053352
23509154
FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(CCC=C)CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions