missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4'-(3-Butenyl)-4-(3,4-difluorophenyl)bicyclohexyl 98.0+%, TCI America™

Catalog No. p-7123836
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49161G 1 g
B49165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49161G Supplier TCI America Supplier No. B49161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 155266-68-5
Molecular Formula C22H30F2
Molecular Weight (g/mol) 332.48
MDL Number MFCD11053352
InChI Key UMULWEQZNFZORL-UHFFFAOYSA-N
PubChem CID 23509154
IUPAC Name 4-(but-3-en-1-yl)-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
SMILES FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(CCC=C)CC1

Specifications

Melting Point 43°C
Color White
Quantity 1 g
Formula Weight 332.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-(3-Butenyl)-4-(3,4-difluorophenyl)bicyclohexyl