missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene 99.0+%, TCI America™

Catalog No. p-7126482
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08795G 5 g
D087925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08795G Supplier TCI America Supplier No. D08795G
Only null left

Chemical Identifiers

CAS 886-65-7
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym 1e,3z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenylbutadiene, e,z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenyl-1,3-butadiene, 1,3-butadiene, 1,4-diphenyl-, e,z, 1,4-diphenyl-1,3-butadiene, 1e,3z, benzene,1'-1,3-butadiene-1,4-diyl bis, 1z,3e-4-phenylbuta-1,3-dienyl benzene, benzene,1,1'-1,3-butadiene-1,4-diyl bis, 1,4-diphenyl-1,3-butadiene e,z-form mi
PubChem CID 1551381
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

Melting Point 152°C
Boiling Point 350°C
Quantity 5 g
Formula Weight 206.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene