accessibility menu, dialog, popup

UserName

(E)(E)

CAS RN 886-65-7

IUPAC Name: [(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
Synonyms: [(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
Molecular Weight (g/mol): 206.29
Molecular Formula: C16H14
InChi Key: JFLKFZNIIQFQBS-FNCQTZNRSA-N
SMILES: C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results