missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (+)-trans-Taxifolin

$417.92 - $1204.61

Chemical Identifiers

CAS 480-18-2
Molecular Formula C15H12O7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC467031000
View Documents
Thermo Scientific Chemicals
467031000
100 mg
Each for $417.92
Only null left
 
AC467035000
View Documents
Thermo Scientific Chemicals
467035000
500 mg
Each for $1,204.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

480-18-2
C15H12O7

Specifications

480-18-2
216.0°C to 220.0°C
304.25
O[C@@H]1[C@H](OC2=CC(O)=CC(O)=C2C1=O)C1=CC=C(O)C(O)=C1
304.25
100 mg
C15H12O7
CXQWRCVTCMQVQX-LSDHHAIUSA-N
(2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
(+)-trans-Taxifolin
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 207-543-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only