missing translation for 'onlineSavingsMsg'
Learn More

trans-Styrylacetic acid, 96%

Catalog No. AC168100010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168100010 1 g Glass bottle
AC168100100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168100010 Supplier Thermo Scientific Chemicals Supplier No. 168100010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1914-58-5 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1914-58-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002783
InChI Key PSCXFXNEYIHJST-QPJJXVBHSA-N
Synonym trans-styrylacetic acid, e-4-phenylbut-3-enoic acid, 3e-4-phenylbut-3-enoic acid, 4-phenyl-3-butenoic acid, 3-butenoic acid, 4-phenyl, 3-butenoic acid, 4-phenyl-, 3e, styrylacetic acid, e-4-phenyl-3-butenoic acid, e-styrylacetic acid
PubChem CID 5370625
SMILES OC(=O)C\C=C\C1=CC=CC=C1

Specifications

Melting Point 82°C to 88°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2CO2H
Quantity 1 g
Beilstein 09, 612
IUPAC Name (3E)-4-phenylbut-3-enoic acid
Formula Weight 162.19
Percent Purity 96%
Physical Form Crystalline Powder or Needles
Chemical Name or Material trans-Styrylacetic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only