accessibility menu, dialog, popup

UserName

CAS RN 1914-58-5

IUPAC Name: (3E)-4-phenylbut-3-enoic acid
Synonyms: 3-butenoic acid, 4-phenyl-
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: PSCXFXNEYIHJST-QPJJXVBHSA-N
SMILES: OC(=O)C\C=C\C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results