missing translation for 'onlineSavingsMsg'
Learn More

trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min

Catalog No. p-7048418
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1049103 1 g
AA1049106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1049103 Supplier Thermo Scientific Chemicals Supplier No. 01049103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation | CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol

As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name ethane;methane;palladium(2+);triphenylphosphane;dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼310°C (decomposition)
Color Yellow
Odor Odorless
Linear Formula PdCl2[P(C6H5)3]2
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Soluble in benzene,and toluene.
Formula Weight 701.91
Concentration or Composition (by Analyte or Components) Pd 14.0% min.
Physical Form Crystalline
Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)
Show More Show Less
EINECSNumber 237-744-2
RTECSNumber RT3578000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only