missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Bromo-beta-nitrostyrene, 99%

Catalog No. AC440160010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440160010 1 g Glass bottle
AC440160050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440160010 Supplier Thermo Scientific Chemicals Supplier No. 440160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5153-71-9 | C8H6BrNO2 | 228.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5153-71-9
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.05
InChI Key LSGVHLGCJIBLMB-AATRIKPKSA-N
Synonym 1-bromo-4-2-nitrovinyl benzene, e-1-bromo-4-2-nitrovinyl benzene, 1-bromo-4-e-2-nitroethenyl benzene, trans-4-bromo-beta-nitrostyrene, benzene, 1-bromo-4-1e-2-nitroethenyl, beta-nitro-4-bromostyrene, 4-bromo-.beta.-nitrostyrene, e-4-bromo-beta-nitrostyrene, 1-bromo-4-e-2-nitrovinyl benzene
PubChem CID 688204
IUPAC Name 1-bromo-4-[(E)-2-nitroethenyl]benzene
SMILES C1=CC(=CC=C1C=C[N+](=O)[O-])Br

Specifications

Melting Point 148.0°C to 152.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 228.05
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material trans-4-Bromo-β-nitrostyrene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only