accessibility menu, dialog, popup

UserName

CAS RN 5153-71-9

IUPAC Name: 1-bromo-4-(2-nitroethenyl)benzene
Synonyms: 1-bromo-4-(2-nitroethenyl)benzene
Molecular Weight (g/mol): 228.05
Molecular Formula: C8H6BrNO2
InChi Key: LSGVHLGCJIBLMB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C=CC1=CC=C(Br)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results