missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-n-Pentylcyclohexyl)benzoic acid, 99%

Catalog No. AAH5735103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5735103 1 g
AAH5735106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5735103 Supplier Thermo Scientific Chemicals Supplier No. H5735103

CAS: 65355-30-8 | C18H26O2 | 274.404 g/mol

It has been used for intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used for intermediate of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 65355-30-8
Molecular Formula C18H26O2
Molecular Weight (g/mol) 274.404
MDL Number MFCD06208360
InChI Key YXKKMVGGPRVHIL-UHFFFAOYSA-N
Synonym 4-4-pentylcyclohexyl benzoic acid, 4-trans-4-n-pentylcyclohexyl benzoic acid, 4-trans-4-pentylcyclohexyl benzoic acid, 4-1s,4r-4-pentylcyclohexyl benzoic acid, 4-pentyl-cyclohexyl benzoic acid, 4-pentyl cyclohexyl benzoic acid, yxkkmvggprvhil-shtzxodssa-n, 4-4-pentyl-cyclohexyl-benzoic acid, 4-trans-4-amylcyclohexyl benzoic acid
PubChem CID 3104716
IUPAC Name 4-(4-pentylcyclohexyl)benzoic acid
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 274.4
Percent Purity 99%
Chemical Name or Material trans-4-(4-n-Pentylcyclohexyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only