accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 65355-30-8

IUPAC Name: 4-(4-pentylcyclohexyl)benzoic acid
Synonyms: 4-(4-pentylcyclohexyl)benzoic acid
Molecular Weight (g/mol): 274.4
Molecular Formula: C18H26O2
InChi Key: YXKKMVGGPRVHIL-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)