Learn More
trans-3-Chloroacrylic acid, 99%
CAS: 2345-61-1 | C3H3ClO2 | 106.505 g/mol
Supplier: Thermo Scientific Chemicals A1552703
Description
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.
Solubility
Soluble in water.
Notes
Store at 4°C. Store away from oxidizing agent, bases.
Specifications
| trans-3-Chloroacrylic acid | |
| 81°C to 85°C | |
| MFCD00064237 | |
| UN3261 | |
| trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546 | |
| MHMUCYJKZUZMNJ-OWOJBTEDSA-N | |
| (E)-3-chloroprop-2-enoic acid | |
| 638124 | |
| 99% |
| 2345-61-1 | |
| C3H3ClO2 | |
| 1 g | |
| 1720279 | |
| Soluble in water. | |
| C(=CCl)C(=O)O | |
| 106.505 | |
| 106.51 |
Chemical Identifiers
| 2345-61-1 | |
| MFCD00064237 | |
| trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546 | |
| (E)-3-chloroprop-2-enoic acid |
| C3H3ClO2 | |
| MHMUCYJKZUZMNJ-OWOJBTEDSA-N | |
| 638124 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 219-070-0
RTECSNumber : AS5960000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only