missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Chloroacrylic acid, 99%

Catalog No. p-7022652
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15527MD 250 mg
AAA1552703 1 g
AAA1552706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15527MD Supplier Thermo Scientific Chemicals Supplier No. A15527MD

CAS: 2345-61-1 | C3H3ClO2 | 106.505 g/mol

trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.

Solubility
Soluble in water.

Notes
Store at 4°C. Store away from oxidizing agent, bases.

Chemical Identifiers

CAS 2345-61-1
Molecular Formula C3H3ClO2
MDL Number MFCD00064237
InChI Key MHMUCYJKZUZMNJ-OWOJBTEDSA-N
Synonym trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546
PubChem CID 638124
IUPAC Name (E)-3-chloroprop-2-enoic acid

Specifications

Melting Point 81°C to 85°C
Quantity 250 mg
UN Number UN3261
Beilstein 1720279
Solubility Information Soluble in water.
SMILES C(=CCl)C(=O)O
Molecular Weight (g/mol) 106.505
Formula Weight 106.51
Percent Purity 99%
Chemical Name or Material trans-3-Chloroacrylic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 219-070-0
RTECSNumber AS5960000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only