accessibility menu, dialog, popup

UserName

CAS RN 2345-61-1

IUPAC Name: (2E)-3-chloroprop-2-enoic acid
Synonyms: trans-3-chloroacrylic acid acrofol
Molecular Weight (g/mol): 106.51
Molecular Formula: C3H3ClO2
InChi Key: MHMUCYJKZUZMNJ-OWOJBTEDSA-N
SMILES: OC(=O)\C=C\Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)