missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-(Boc-amino)cyclobutanemethanol, 97%
CAS: 167081-37-0 | C10H19NO3 | 201.27 g/mol
Supplier: Thermo Scientific Chemicals H6271903
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 167081-37-0 | |
| 201.27 | |
| PCPNTJQMXAHNOA-UHFFFAOYSA-N | |
| 22408683 | |
| CC(C)(C)OC(=O)NC1CC(CO)C1 |
| C10H19NO3 | |
| MFCD08166748,MFCD08436074,MFCD09864183 | |
| tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol | |
| tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate |
Specifications
| trans-3-(Boc-amino)cyclobutanemethanol | |
| C10H19NO3 | |
| tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol | |
| CC(C)(C)OC(=O)NC1CC(CO)C1 | |
| 1 g | |
| 22408683 | |
| 97% |
| 167081-37-0 | |
| MFCD08166748,MFCD08436074,MFCD09864183 | |
| PCPNTJQMXAHNOA-UHFFFAOYSA-N | |
| tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate | |
| 201.27 | |
| 201.26 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures