missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclobutanemethanol, 97%

Catalog No. p-7041588
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62719MD 250 mg
AAH6271903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62719MD Supplier Thermo Scientific Chemicals Supplier No. H62719MD

CAS: 167081-37-0 | C10H19NO3 | 201.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167081-37-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.27
MDL Number MFCD08166748,MFCD08436074,MFCD09864183
InChI Key PCPNTJQMXAHNOA-UHFFFAOYSA-N
Synonym tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol
PubChem CID 22408683
IUPAC Name tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CC(CO)C1

Specifications

Quantity 250 mg
Formula Weight 201.26
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclobutanemethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only