accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOH

CAS RN 167081-37-0

IUPAC Name: tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate
Synonyms: tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate
Molecular Weight (g/mol): 201.27
Molecular Formula: C10H19NO3
InChi Key: PCPNTJQMXAHNOA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CC(CO)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)