missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol, 97%, Thermo Scientific™

$889.71 - $889.71

Chemical Identifiers

CAS 1202411-95-7
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.337
MDL Number MFCD09864989
InChI Key QYHAKPLUUUMEFH-ZIAGYGMSSA-N
Synonym benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
PubChem CID 59401466
IUPAC Name benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6284203
View Documents
Thermo Scientific Chemicals
H6284203
1 g
Each for $889.71
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1202411-95-7
263.337
QYHAKPLUUUMEFH-ZIAGYGMSSA-N
59401466
C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO
C15H21NO3
MFCD09864989
benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate

Specifications

1202411-95-7
MFCD09864989
QYHAKPLUUUMEFH-ZIAGYGMSSA-N
benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
263.337
263.33
trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol
C15H21NO3
benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO
1 g
59401466
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents