accessibility menu, dialog, popup

UserName

CAS RN 1202411-95-7

HNOOHO

CAS RN 1202411-95-7

IUPAC Name: benzyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Synonyms: benzyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 263.34
Molecular Formula: C15H21NO3
InChi Key: QYHAKPLUUUMEFH-UHFFFAOYNA-N
SMILES: OCC1CCCC(C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results