missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6284203
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol | |
| C15H21NO3 | |
| benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate | |
| C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO | |
| 1 g | |
| 59401466 | |
| 97% |
| 1202411-95-7 | |
| MFCD09864989 | |
| QYHAKPLUUUMEFH-ZIAGYGMSSA-N | |
| benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate | |
| 263.337 | |
| 263.33 |
Chemical Identifiers
| 1202411-95-7 | |
| 263.337 | |
| QYHAKPLUUUMEFH-ZIAGYGMSSA-N | |
| 59401466 | |
| C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO |
| C15H21NO3 | |
| MFCD09864989 | |
| benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate | |
| benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures