Learn More
trans-3-(3-Pyridyl)acrylic acid, 99%
CAS: 19337-97-4 | C8H7NO2 | 149.149 g/mol
Supplier: Thermo Scientific Chemicals A1474714
Description
trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.
Notes
Incompatible with oxidizing agents, acids and bases.
Specifications
| trans-3-(3-Pyridyl)acrylic acid | |
| 232°C to 235°C | |
| MFCD00006410 | |
| 113290 | |
| VUVORVXMOLQFMO-ONEGZZNKSA-N | |
| (E)-3-pyridin-3-ylprop-2-enoic acid | |
| 776396 | |
| 99% |
| 19337-97-4 | |
| C8H7NO2 | |
| 25 g | |
| 3-3-pyridyl acrylic acid, trans-3-3-pyridyl acrylic acid, e-3-pyridin-3-yl acrylic acid, 3-pyridylacrylic acid, 2e-3-pyridin-3-ylacrylic acid, 2e-3-pyridin-3-yl prop-2-enoic acid, 3-pyridin-3-yl acrylic acid, trans-3-3'-pyridyl acrylic acid, 3-3-pyridinyl acrylic acid, 3-pyridinyl-acrylic acid | |
| C1=CC(=CN=C1)C=CC(=O)O | |
| 149.149 | |
| 149.15 |
Chemical Identifiers
| 19337-97-4 | |
| 149.149 | |
| VUVORVXMOLQFMO-ONEGZZNKSA-N | |
| 776396 | |
| C1=CC(=CN=C1)C=CC(=O)O |
| C8H7NO2 | |
| MFCD00006410 | |
| 3-3-pyridyl acrylic acid, trans-3-3-pyridyl acrylic acid, e-3-pyridin-3-yl acrylic acid, 3-pyridylacrylic acid, 2e-3-pyridin-3-ylacrylic acid, 2e-3-pyridin-3-yl prop-2-enoic acid, 3-pyridin-3-yl acrylic acid, trans-3-3'-pyridyl acrylic acid, 3-3-pyridinyl acrylic acid, 3-pyridinyl-acrylic acid | |
| (E)-3-pyridin-3-ylprop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-424-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only