accessibility menu, dialog, popup

UserName

CAS RN 19337-97-4

IUPAC Name: (2E)-3-(pyridin-3-yl)prop-2-enoic acid
Synonyms: (2E)-3-(pyridin-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: VUVORVXMOLQFMO-ONEGZZNKSA-N
SMILES: OC(=O)\C=C\C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results