missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(3-Pyridyl)acrylic acid, 99%

Catalog No. p-7022125
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1474706 5 g
AAA1474714 25 g
AAA1474730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1474706 Supplier Thermo Scientific Chemicals Supplier No. A1474706

CAS: 19337-97-4 | C8H7NO2 | 149.149 g/mol

trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

Notes
Incompatible with oxidizing agents, acids and bases.

Chemical Identifiers

CAS 19337-97-4
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00006410
InChI Key VUVORVXMOLQFMO-ONEGZZNKSA-N
Synonym 3-3-pyridyl acrylic acid, trans-3-3-pyridyl acrylic acid, e-3-pyridin-3-yl acrylic acid, 3-pyridylacrylic acid, 2e-3-pyridin-3-ylacrylic acid, 2e-3-pyridin-3-yl prop-2-enoic acid, 3-pyridin-3-yl acrylic acid, trans-3-3'-pyridyl acrylic acid, 3-3-pyridinyl acrylic acid, 3-pyridinyl-acrylic acid
PubChem CID 776396
IUPAC Name (E)-3-pyridin-3-ylprop-2-enoic acid
SMILES C1=CC(=CN=C1)C=CC(=O)O

Specifications

Melting Point 232°C to 235°C
Quantity 5 g
Beilstein 113290
Formula Weight 149.15
Percent Purity 99%
Chemical Name or Material trans-3-(3-Pyridyl)acrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-424-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only